Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atg14
Entrez Gene ID
100504663
Gene Name
autophagy related 14
Gene Aliases
4832427M01, Atg14L, D14Ertd114e, D14Ertd436e
Chromosome Location
Chr.14: 47540893 - 47568434 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172599.4 NP_766187.1 1 - 2 252 132
GenBank mRNA AK029330.1 - 1 - 2 233 132
AK029967.1 - 1 - 2 233 132
AK054196.1 - 1 - 2 253 132
BC090995.1 - 1 - 2 232 132

View Full Product Details