Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cgrrf1
Entrez Gene ID
68755
Gene Name
cell growth regulator with ring finger domain 1
Gene Aliases
1110038G02Rik, 1810009H17Rik, CGR19
Chromosome Location
Chr.14: 46832093 - 46854200 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026832.3 NP_081108.2 4 - 5 604 89
GenBank mRNA AK030755.1 - 4 - 5 606 89
AK154797.1 - 4 - 5 680 89
BC037677.1 - 4 - 5 620 89
BC114536.1 - 3 - 4 442 89

View Full Product Details