Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Park2
Entrez Gene ID
50873
Gene Name
Parkinson disease (autosomal recessive, juvenile) 2, parkin
Gene Aliases
PRKN
Chromosome Location
Chr.17: 10840384 - 12063370 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016694.3 NP_057903.1 4 - 5 634 69
GenBank mRNA AB019558.1 - 4 - 5 605 69
AF250293.1 - 4 - 5 634 69
AF250294.1 - 4 - 5 637 69
AF250295.1 - 4 - 5 637 69
BC113204.1 - 4 - 5 617 69
FJ555554.1 - 2 - 3 387 69
FJ555556.1 - 3 - 4 367 69
FJ555557.1 - 4 - 5 608 69

View Full Product Details