Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppm1n
Entrez Gene ID
232941
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1N (putative)
Gene Aliases
A930002H20, C79127
Chromosome Location
Chr.7: 19276807 - 19280194 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177691.3 NP_808359.1 2 - 3 1059 84
GenBank mRNA AK044232.1 - 2 - 3 1059 84
AK044334.1 - 2 - 3 1060 84
BC096372.1 - 2 - 3 1048 84

View Full Product Details