The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166064.1 | NP_001159536.1 | 3 - 4 | 3036 | 71 |
GenBank mRNA | CN702275.1 | - | 1 - 2 | 92 | 71 |