The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011076.2 | NP_035206.2 | 15 - 16 | 2016 | 158 |
GenBank mRNA | AY864315.1 | - | 14 - 15 | 1879 | 158 |
M24417.1 | - | 9 - 10 | 1365 | 158 | |
M30697.1 | - | 15 - 16 | 2030 | 158 | |
M33581.1 | - | 15 - 16 | 2015 | 158 |