The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013886.4 | NP_038914.2 | 5 - 6 | 1040 | 80 |
GenBank mRNA | AK014849.1 | - | 5 - 6 | 864 | 80 |
AK019487.1 | - | 5 - 6 | 703 | 80 | |
AK042303.1 | - | 1 - 2 | 98 | 80 | |
AK044858.1 | - | 5 - 6 | 865 | 80 | |
AK045542.1 | - | 5 - 6 | 1039 | 80 | |
AK162418.1 | - | 5 - 6 | 683 | 80 | |
BC055734.1 | - | 5 - 6 | 627 | 80 |