The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301737.1 | NP_001288666.1 | 20 - 21 | 2441 | 139 |
NM_010065.3 | NP_034195.2 | 19 - 20 | 2429 | 139 | |
NR_125959.1 | 20 - 21 | 2441 | 139 | ||
GenBank mRNA | AK144142.1 | - | 19 - 20 | 2380 | 139 |
AK220483.1 | - | 20 - 21 | 2383 | 139 | |
BC034679.1 | - | 20 - 21 | 2395 | 139 | |
BC058623.1 | - | 20 - 21 | 2375 | 139 | |
L29457.1 | - | 4 - 5 | 467 | 139 | |
L31396.1 | - | 15 - 16 | 1610 | 139 | |
L31397.1 | - | 19 - 20 | 2382 | 139 |