The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252580.1 | NP_001239509.1 | 7 - 8 | 952 | 89 |
NM_019729.3 | NP_062703.2 | 7 - 8 | 919 | 89 | |
GenBank mRNA | AB045709.1 | - | 6 - 7 | 693 | 89 |
AF057146.1 | - | 6 - 7 | 710 | 89 | |
AK122195.1 | - | 7 - 8 | 830 | 89 | |
AK129003.1 | - | 7 - 8 | 920 | 89 | |
AK134058.1 | - | 7 - 8 | 874 | 89 | |
AK142881.1 | - | 4 - 5 | 417 | 89 | |
AK153015.1 | - | 6 - 7 | 667 | 89 | |
AK163469.1 | - | 7 - 8 | 852 | 89 | |
AK166540.1 | - | 7 - 8 | 920 | 89 | |
BC038123.1 | - | 7 - 8 | 904 | 89 | |
BC050947.1 | - | 7 - 8 | 884 | 89 | |
BC061465.1 | - | 7 - 8 | 884 | 89 | |
BC066126.1 | - | 7 - 8 | 882 | 89 |