Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Usp8
Entrez Gene ID
84092
Gene Name
ubiquitin specific peptidase 8
Gene Aliases
AI574262, AW557536, Ubpy, mKIAA0055
Chromosome Location
Chr.2: 126707202 - 126759314 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252580.1 NP_001239509.1 8 - 9 1110 75
NM_019729.3 NP_062703.2 8 - 9 1077 75
GenBank mRNA AB045709.1 - 7 - 8 851 75
AF057146.1 - 7 - 8 868 75
AK122195.1 - 8 - 9 988 75
AK129003.1 - 8 - 9 1078 75
AK134058.1 - 8 - 9 1032 75
AK142881.1 - 5 - 6 575 75
AK153015.1 - 7 - 8 825 75
AK163469.1 - 8 - 9 1010 75
BC038123.1 - 8 - 9 1062 75
BC050947.1 - 8 - 9 1042 75
BC061465.1 - 8 - 9 1042 75
BC066126.1 - 8 - 9 1040 75

View Full Product Details