Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Meis2
Entrez Gene ID
17536
Gene Name
Meis homeobox 2
Gene Aliases
A430109D20Rik, Mrg1, Stra10
Chromosome Location
Chr.2: 115861264 - 116067555 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136072.2 NP_001129544.1 3 - 4 631 90
NM_001159567.1 NP_001153039.1 3 - 4 631 90
NM_001159568.1 NP_001153040.1 3 - 4 631 90
NM_001159569.1 NP_001153041.1 3 - 4 904 90
NM_001159570.1 NP_001153042.1 3 - 4 904 90
NM_010825.3 NP_034955.1 3 - 4 631 90
GenBank mRNA AJ000504.1 - 3 - 4 620 90
AJ000505.1 - 3 - 4 620 90
AJ000506.1 - 3 - 4 620 90
AJ000507.1 - 3 - 4 620 90
AK040611.1 - 1 - 2 912 90
AK045126.1 - 1 - 2 376 90
AK146633.1 - 3 - 4 631 90
AK158513.1 - 3 - 4 796 90
AK158839.1 - 3 - 4 903 90
BC017375.1 - 3 - 4 697 90
KM974644.1 - 1 - 2 114 90
U57343.1 - - 668 90
U68383.1 - 3 - 4 590 90
U68384.1 - 3 - 4 590 90

View Full Product Details