Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gtpbp3
Entrez Gene ID
70359
Gene Name
GTP binding protein 3
Gene Aliases
2410009F13Rik, AI607903
Chromosome Location
Chr.8: 71487870 - 71499623 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032544.3 NP_115933.2 7 - 8 1666 119
GenBank mRNA AK046330.1 - 7 - 8 1042 119
BC055716.1 - 7 - 8 1258 119
BC075712.1 - 4 - 5 1696 119
BC138560.1 - 7 - 8 1075 119

View Full Product Details