Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r28
Entrez Gene ID
665255
Gene Name
vomeronasal 2, receptor 28
Gene Aliases
EG665255
Chromosome Location
Chr.7: 5480456 - 5493851 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081405.1 NP_001074874.1 1 - 2 196 95

View Full Product Details