The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177869.1 | NP_001171340.1 | 11 - 12 | 5037 | 64 |
NR_033218.1 | 11 - 12 | 6336 | 64 | ||
NR_033615.1 | 9 - 10 | 3031 | 64 | ||
GenBank mRNA | AK147260.1 | - | 11 - 12 | 6337 | 64 |
AK147653.1 | - | 11 - 12 | 6337 | 64 | |
AY946044.1 | - | 9 - 10 | 4728 | 64 | |
BC027834.1 | - | 1 - 2 | 591 | 64 |