The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026830.2 | NP_081106.1 | 1 - 2 | 307 | 107 |
GenBank mRNA | AK154980.1 | - | 1 - 2 | 307 | 107 |
BC080680.1 | - | 1 - 2 | 153 | 107 | |
BY206933.1 | - | 1 - 2 | 102 | 107 |