The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039188.1 | NP_001034277.1 | 5 - 6 | 854 | 86 |
NM_001177868.1 | NP_001171339.1 | 5 - 6 | 579 | 86 | |
NM_001177869.1 | NP_001171340.1 | 4 - 5 | 515 | 86 | |
NM_026830.2 | NP_081106.1 | 5 - 6 | 822 | 86 | |
NR_033218.1 | 5 - 6 | 1976 | 86 | ||
NR_033615.1 | 5 - 6 | 1793 | 86 | ||
GenBank mRNA | AK047047.1 | - | 5 - 6 | 1773 | 86 |
AK047251.1 | - | 5 - 6 | 1775 | 86 | |
AK147260.1 | - | 5 - 6 | 1977 | 86 | |
AK147653.1 | - | 5 - 6 | 1977 | 86 | |
AK154980.1 | - | 5 - 6 | 822 | 86 | |
AK170652.1 | - | 5 - 6 | 854 | 86 | |
AK171375.1 | - | 5 - 6 | 559 | 86 | |
AY946044.1 | - | 2 - 3 | 206 | 86 | |
BC080680.1 | - | 5 - 6 | 668 | 86 |