The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009832.2 | NP_033962.2 | 2 - 3 | 177 | 109 |
GenBank mRNA | AK089386.1 | - | 2 - 3 | 114 | 109 |
AK146711.1 | - | 2 - 3 | 114 | 109 | |
AK154679.1 | - | 2 - 3 | 114 | 109 | |
AK154765.1 | - | 2 - 3 | 114 | 109 | |
BC027297.1 | - | 2 - 3 | 72 | 109 |