Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmc4
Entrez Gene ID
353499
Gene Name
transmembrane channel-like gene family 4
Gene Aliases
-
Chromosome Location
Chr.7: 3665754 - 3677553 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181820.2 NP_861541.2 6 - 7 983 58
GenBank mRNA AK050105.1 - 5 - 6 878 58
AY263162.1 - 6 - 7 954 58
BC040466.1 - 6 - 7 895 58
BC055939.1 - 5 - 6 712 58

View Full Product Details