Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpvl
Entrez Gene ID
71287
Gene Name
carboxypeptidase, vitellogenic-like
Gene Aliases
4933436L16Rik, HVLP
Chromosome Location
Chr.6: 53873279 - 53978673 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289713.1 NP_001276642.1 11 - 12 1189 65
NM_001289714.1 NP_001276643.1 10 - 11 1057 65
NM_027749.2 NP_082025.1 11 - 12 1275 65
GenBank mRNA AK017087.1 - 10 - 11 1058 65
BC137839.1 - 11 - 12 1264 65
BC144966.1 - 11 - 12 1178 65

View Full Product Details