Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmc1
Entrez Gene ID
19179
Gene Name
protease (prosome, macropain) 26S subunit, ATPase 1
Gene Aliases
AI325227, P26s4, Rpt2/S4, S4, rpt2
Chromosome Location
Chr.12: 100110202 - 100123364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK088411.1 - 1 - 2 40 114
AK133926.1 - 1 - 2 44 114
AK161641.1 - 1 - 2 59 114
AK161723.1 - 1 - 2 60 114

View Full Product Details