Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdcl
Entrez Gene ID
67466
Gene Name
phosducin-like
Gene Aliases
1200011E13Rik, AW108059, PhLP1
Chromosome Location
Chr.2: 37350074 - 37359820 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK004704.1 - 1 - 2 901 63

View Full Product Details