Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ehmt1
Entrez Gene ID
77683
Gene Name
euchromatic histone methyltransferase 1
Gene Aliases
9230102N17Rik, D330003E03, Eu-HMTase1, GLP, GLP1, KMT1D, mKIAA1876
Chromosome Location
Chr.2: 24790769 - 24919643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012518.3 NP_001012536.2 3 - 4 676 123
NM_001109686.2 NP_001103156.1 3 - 4 676 123
NM_001109687.2 NP_001103157.1 3 - 4 676 123
NM_172545.4 NP_766133.2 3 - 4 676 123
GenBank mRNA AB205007.1 - 3 - 4 682 123
AK043898.1 - 3 - 4 672 123
AK052174.1 - 3 - 4 659 123
AK054555.1 - 3 - 4 644 123
AK082062.1 - 3 - 4 666 123
AK084994.1 - 3 - 4 659 123
AK086833.1 - 1 - 2 562 123
BC056938.1 - 1 - 2 377 123
BC089302.1 - 1 - 2 544 123

View Full Product Details