The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139504.1 | NP_001132976.1 | 2 - 3 | 487 | 69 |
NM_026828.3 | NP_081104.1 | 2 - 3 | 483 | 69 | |
GenBank mRNA | AK004089.1 | - | 2 - 3 | 473 | 69 |
AK142070.1 | - | 2 - 3 | 484 | 69 | |
BC139333.1 | - | 2 - 3 | 447 | 69 | |
BC139334.1 | - | 2 - 3 | 446 | 69 |