The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081086.1 | NP_001074555.1 | 3 - 4 | 429 | 95 |
GenBank mRNA | AK088007.1 | - | 3 - 4 | 240 | 95 |
AK134121.1 | - | 3 - 4 | 431 | 96 | |
AK135178.1 | - | 4 - 5 | 561 | 95 | |
BC016126.1 | - | 3 - 4 | 291 | 95 | |
BC027420.1 | - | 3 - 4 | 423 | 95 | |
BC038818.1 | - | 3 - 4 | 247 | 95 | |
BC048905.1 | - | 3 - 4 | 290 | 95 | |
BC094926.1 | - | 3 - 4 | 283 | 95 | |
BC150694.1 | - | 4 - 5 | 450 | 95 |