Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fastkd1
Entrez Gene ID
320720
Gene Name
FAST kinase domains 1
Gene Aliases
5330408N05Rik, mKIAA1800
Chromosome Location
Chr.2: 69686820 - 69712606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177244.3 NP_796218.2 9 - 10 2045 68
GenBank mRNA AK220423.1 - 5 - 6 893 68
BC023501.1 - 2 - 3 178 68
BC034292.1 - 6 - 7 1152 68
BC069943.1 - 8 - 9 1538 68
BC075689.1 - 9 - 10 2045 68

View Full Product Details