Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hhipl1
Entrez Gene ID
214305
Gene Name
hedgehog interacting protein-like 1
Gene Aliases
1600002O04Rik, AK129450, mKIAA1822
Chromosome Location
Chr.12: 108305610 - 108328300 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044380.1 NP_001037845.1 4 - 5 1534 70
GenBank mRNA AK129450.1 - - 929 70
AK138646.1 - 4 - 5 1258 70
AK147996.1 - - 1201 70
AK166269.1 - - 1430 70
BC057108.1 - - 527 70
BC113163.1 - 4 - 5 1534 70
BC113773.1 - 4 - 5 1530 70

View Full Product Details