Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Myh8
- Entrez Gene ID
-
17885
- Gene Name
- myosin, heavy polypeptide 8, skeletal muscle, perinatal
- Gene Aliases
-
4832426G23Rik, AI327267, MHCp, MyHC-pn, Myhs-p, Myhsp
- Chromosome Location
-
Chr.11: 67277124 - 67308634 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_177369.3 |
NP_796343.2 |
38 - 39 |
5757 |
96 |
View Full Product Details