The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001093764.1 | NP_001087233.1 | 2 - 3 | 378 | 64 |
NM_001093765.1 | NP_001087234.1 | 2 - 3 | 327 | 64 | |
NM_001093766.1 | NP_001087235.1 | 1 - 2 | 265 | 64 | |
NM_016969.2 | NP_058665.2 | 2 - 3 | 166 | 64 | |
GenBank mRNA | AJ001616.1 | - | 2 - 3 | 187 | 64 |
AK052258.1 | - | 2 - 3 | 285 | 64 | |
AK053414.1 | - | 2 - 3 | 118 | 64 | |
AK089538.1 | - | 2 - 3 | 149 | 64 | |
AK149962.1 | - | 2 - 3 | 309 | 64 | |
AK159160.1 | - | - | 309 | 64 | |
BC056355.1 | - | 2 - 3 | 261 | 64 | |
BC119364.1 | - | 1 - 2 | 49 | 64 | |
BC119368.1 | - | 1 - 2 | 46 | 64 | |
BY252968.1 | - | 2 - 3 | 320 | 64 | |
CF166357.1 | - | 1 - 2 | 265 | 64 | |
CK782239.1 | - | 2 - 3 | 367 | 64 |