Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eng
Entrez Gene ID
13805
Gene Name
endoglin
Gene Aliases
AI528660, AI662476, CD105, Endo, S-endoglin
Chromosome Location
Chr.2: 32646532 - 32682669 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146348.1 NP_001139820.1 7 - 8 1350 90
NM_001146350.1 NP_001139822.1 7 - 8 1353 90
NM_007932.2 NP_031958.2 7 - 8 1353 90
GenBank mRNA AK151302.1 - 7 - 8 1323 90
AY679531.1 - 7 - 8 1353 90
BC029080.1 - 7 - 8 1267 90
S69407.1 - 8 - 9 1218 90
X77952.1 - 7 - 8 1353 90

View Full Product Details