Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map1a
Entrez Gene ID
17754
Gene Name
microtubule-associated protein 1 A
Gene Aliases
6330416M19Rik, AI853608, Mtap-1, Mtap1, Mtap1a, moth1
Chromosome Location
Chr.2: 121289600 - 121310832 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173506.1 NP_001166977.1 2 - 3 170 60
NM_032393.2 NP_115769.1 3 - 4 454 60
GenBank mRNA AF182212.1 - 2 - 3 161 60
AF182213.1 - 3 - 4 454 60
AK160546.1 - 2 - 3 154 60
BC028853.1 - 2 - 3 160 60
CJ116735.1 - 2 - 3 171 60

View Full Product Details