Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lcn9
Entrez Gene ID
77704
Gene Name
lipocalin 9
Gene Aliases
9230102I19Rik
Chromosome Location
Chr.2: 25823094 - 25825542 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029959.2 NP_084235.1 6 - 7 573 117
GenBank mRNA AF435738.1 - 6 - 7 573 117
AK020305.1 - 6 - 7 575 117

View Full Product Details