The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029293.2 | NP_083569.1 | 1 - 2 | 195 | 58 |
GenBank mRNA | AK005756.1 | - | 1 - 2 | 195 | 58 |
BC028657.1 | - | 1 - 2 | 168 | 58 |