Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nudt14
Entrez Gene ID
66174
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 14
Gene Aliases
1110030M18Rik
Chromosome Location
Chr.12: 112934727 - 112942167 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025399.3 NP_079675.1 4 - 5 473 66
GenBank mRNA AK003991.1 - 4 - 5 480 66
AK013174.1 - 4 - 5 429 66
BC025444.1 - 4 - 5 475 66

View Full Product Details