Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efcab11
Entrez Gene ID
78767
Gene Name
EF-hand calcium binding domain 11
Gene Aliases
2610021K21Rik, 2610110A13Rik
Chromosome Location
Chr.12: 99692752 - 99884445 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030172.2 NP_084448.1 4 - 5 399 99
GenBank mRNA AK011507.1 - 4 - 5 399 99
AK011836.1 - 4 - 5 381 99
AK035051.1 - 4 - 5 379 99
BC049109.1 - 4 - 5 361 99

View Full Product Details