The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030172.2 | NP_084448.1 | 5 - 6 | 497 | 65 |
GenBank mRNA | AK011507.1 | - | 5 - 6 | 497 | 65 |
AK035051.1 | - | 5 - 6 | 477 | 65 | |
BC049109.1 | - | 5 - 6 | 459 | 65 |