Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gtf2h5
- Entrez Gene ID
-
66467
- Gene Name
- general transcription factor IIH, polypeptide 5
- Gene Aliases
-
2700017P07Rik, 2810432H05Rik, D17Wsu155e
- Chromosome Location
-
Chr.17: 6079828 - 6085488 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK012257.1 |
- |
1 - 2 |
149 |
81 |
View Full Product Details