Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chrdl1
Entrez Gene ID
83453
Gene Name
chordin-like 1
Gene Aliases
CHL, CHL1, Nrln1, VOPT
Chromosome Location
Chr.X: 143285674 - 143394262 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114385.1 NP_001107857.1 8 - 9 1287 93
NM_031258.3 NP_112548.2 8 - 9 1287 93
GenBank mRNA AF296451.1 - 8 - 9 1209 93
AF305714.1 - 7 - 8 826 93
AF321853.1 - 7 - 8 763 93
AK142805.1 - 8 - 9 911 93
AK164661.1 - 8 - 9 882 93
BC050818.1 - 7 - 8 777 93
BC066832.1 - 8 - 9 921 93

View Full Product Details