Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rxra
Entrez Gene ID
20181
Gene Name
retinoid X receptor alpha
Gene Aliases
9530071D11Rik, Nr2b1, RXRalpha1
Chromosome Location
Chr.2: 27676324 - 27763319 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290481.1 NP_001277410.1 8 - 9 1197 64
NM_001290482.1 NP_001277411.1 8 - 9 1234 64
NM_011305.3 NP_035435.1 8 - 9 1376 64
GenBank mRNA AK031034.1 - 8 - 9 1245 64
AK144675.1 - 8 - 9 1378 64
BC138800.1 - 9 - 10 1227 64
BC138802.1 - 9 - 10 1227 64
M84817.1 - 9 - 10 1239 64
U77683.1 - 8 - 9 1207 64
U77684.1 - 8 - 9 1237 64
X66223.1 - 8 - 9 1321 64

View Full Product Details