Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stom
Entrez Gene ID
13830
Gene Name
stomatin
Gene Aliases
Epb7.2
Chromosome Location
Chr.2: 35313990 - 35337009 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013515.2 NP_038543.1 2 - 3 251 109
GenBank mRNA AF093620.1 - 2 - 3 202 109
AK077435.1 - 2 - 3 219 109
AK139242.1 - 2 - 3 219 109
AK143724.1 - 2 - 3 219 109
AK148975.1 - 2 - 3 220 109
AK149689.1 - 2 - 3 219 109
AK149821.1 - 2 - 3 219 109
AK149991.1 - 2 - 3 219 109
AK151129.1 - 2 - 3 218 109
BC003789.1 - 2 - 3 214 109
U17297.1 - 2 - 3 251 109

View Full Product Details