The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163434.1 | NP_001156906.1 | 6 - 7 | 1146 | 68 |
NM_022310.3 | NP_071705.3 | 5 - 6 | 1228 | 68 | |
GenBank mRNA | AJ002387.1 | - | 5 - 6 | 1007 | 68 |
AK004578.1 | - | 5 - 6 | 1223 | 68 | |
AK076079.1 | - | 6 - 7 | 1142 | 68 | |
AK077463.1 | - | 6 - 7 | 1147 | 68 | |
AK144650.1 | - | 6 - 7 | 1143 | 68 | |
AK146647.1 | - | 6 - 7 | 1145 | 68 | |
AK148539.1 | - | 6 - 7 | 1145 | 68 | |
AK151244.1 | - | 5 - 6 | 1095 | 68 | |
AK151647.1 | - | 6 - 7 | 1144 | 68 | |
AK151806.1 | - | 6 - 7 | 1145 | 68 | |
AK152020.1 | - | 6 - 7 | 1142 | 68 | |
AK152521.1 | - | 5 - 6 | 1225 | 68 | |
AK152958.1 | - | 6 - 7 | 1145 | 68 | |
AK159719.1 | - | 6 - 7 | 1142 | 68 | |
AK160592.1 | - | 1 - 2 | 146 | 68 | |
AK160821.1 | - | - | 1145 | 68 | |
AK166207.1 | - | 6 - 7 | 1142 | 68 | |
AK166739.1 | - | 6 - 7 | 1142 | 68 | |
AK167011.1 | - | 6 - 7 | 1142 | 68 | |
AK168012.1 | - | 6 - 7 | 1146 | 68 | |
AK168230.1 | - | 6 - 7 | 1140 | 68 | |
AK169034.1 | - | 6 - 7 | 1146 | 68 | |
BC005785.1 | - | 4 - 5 | 862 | 68 | |
BC050927.1 | - | 5 - 6 | 1205 | 68 |