Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Swi5
Entrez Gene ID
72931
Gene Name
SWI5 recombination repair homolog (yeast)
Gene Aliases
1500019F05Rik, 2900010J23Rik, AI046671
Chromosome Location
Chr.2: 32278805 - 32288136 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_110965.1 2 - 3 233 77
GenBank mRNA AK018775.1 - 2 - 3 171 77

View Full Product Details