The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039653.2 | NP_001034742.1 | 1 - 2 | 237 | 94 |
GenBank mRNA | BC132554.1 | - | 1 - 2 | 151 | 94 |
BC132556.1 | - | 1 - 2 | 151 | 94 | |
BY716979.1 | - | 1 - 2 | 218 | 94 | |
L38248.1 | - | 1 - 2 | 203 | 94 |