The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026044.3 | NP_080320.1 | 8 - 9 | 1000 | 82 |
GenBank mRNA | AK013297.1 | - | 8 - 9 | 980 | 82 |
AK078448.1 | - | 8 - 9 | 963 | 82 | |
AK152000.1 | - | 7 - 8 | 913 | 82 | |
AK153537.1 | - | 7 - 8 | 913 | 82 | |
BC056962.1 | - | 6 - 7 | 1650 | 82 | |
BC086486.1 | - | 8 - 9 | 1002 | 82 |