Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atg13
Entrez Gene ID
51897
Gene Name
autophagy related 13
Gene Aliases
1110053A20Rik, D2Ertd391e, Harbi1
Chromosome Location
Chr.2: 91674612 - 91710598 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145528.3 NP_663503.1 2 - 3 425 73
GenBank mRNA AK149672.1 - 2 - 3 384 73
AK154470.1 - 2 - 3 411 73
AK163962.1 - 2 - 3 377 73
AK171206.1 - 2 - 3 411 73
AK171440.1 - 2 - 3 402 73
BC016669.1 - 2 - 3 409 73
BC023673.1 - 2 - 3 375 73
BC023712.1 - 2 - 3 398 73
BC033419.1 - 2 - 3 376 73

View Full Product Details