The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007656.5 | NP_031682.1 | 1 - 2 | 167 | 83 |
GenBank mRNA | AK152871.1 | - | 1 - 2 | 162 | 83 |
AK153070.1 | - | 1 - 2 | 162 | 83 | |
AK158033.1 | - | 1 - 2 | 110 | 83 | |
AK168515.1 | - | 1 - 2 | 137 | 83 | |
BC132559.1 | - | 1 - 2 | 137 | 83 | |
BC132565.1 | - | 1 - 2 | 137 | 83 |