Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olfr74
Entrez Gene ID
117005
Gene Name
olfactory receptor 74
Gene Aliases
MOR174-4, mOR-EV
Chromosome Location
Chr.2: 87973707 - 87974663 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054091.2 NP_473432.2 1 - 1 541 94
GenBank mRNA AB061229.1 - 1 - 1 541 94

View Full Product Details