Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gtpbp3
Entrez Gene ID
70359
Gene Name
GTP binding protein 3
Gene Aliases
2410009F13Rik, AI607903
Chromosome Location
Chr.8: 71487870 - 71499623 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032544.3 NP_115933.2 1 - 2 741 105
GenBank mRNA AF361482.2 - 1 - 2 741 105
AK046330.1 - 1 - 2 117 105
BC055716.1 - 1 - 2 93 105
BC075712.1 - 1 - 2 101 105
BC092042.1 - 1 - 2 124 105
BC138560.1 - 1 - 2 150 105
BC145333.1 - 1 - 2 150 105

View Full Product Details