The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159640.2 | NP_001153112.1 | 4 - 5 | 399 | 119 |
NM_001159641.2 | NP_001153113.1 | 4 - 5 | 399 | 119 | |
NM_001289611.1 | NP_001276540.1 | 4 - 5 | 399 | 119 | |
NM_175225.5 | NP_780434.1 | 4 - 5 | 666 | 119 | |
NR_110350.1 | 4 - 5 | 666 | 119 | ||
NR_110351.1 | 4 - 5 | 399 | 119 | ||
GenBank mRNA | AK015624.1 | - | 4 - 5 | 399 | 119 |
AK036512.1 | - | 4 - 5 | 622 | 119 | |
AK050603.1 | - | 1 - 2 | 123 | 119 | |
AK146320.1 | - | 4 - 5 | 382 | 119 | |
AK160983.1 | - | 4 - 5 | 627 | 119 | |
BB665115.1 | - | 4 - 5 | 375 | 119 | |
BC024597.1 | - | 4 - 5 | 386 | 119 |