The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123327.2 | NP_001116799.1 | 11 - 12 | 5169 | 157 |
GenBank mRNA | AK141733.1 | - | 9 - 10 | 3480 | 157 |