Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tekt3
Entrez Gene ID
71062
Gene Name
tektin 3
Gene Aliases
4933407G07Rik
Chromosome Location
Chr.11: 63061659 - 63094960 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027660.1 NP_081936.1 4 - 5 884 70
GenBank mRNA AY252101.1 - 4 - 5 884 70
BC127605.1 - 4 - 5 782 70
BC128270.1 - 4 - 5 782 70
BC139074.1 - 4 - 5 851 70
BC139075.1 - 4 - 5 851 70

View Full Product Details